2-Oxazolidinone 98%

Supplier: Thermo Scientific Chemicals

12966-0050 12966-0250 12966-1000
TS12966-0050EA 23 USD
TS12966-0050 TS12966-0250 TS12966-1000
2-Oxazolidinone 98%
2-Oxazolidinone
Formula: C₃H₅NO₂
Boiling Pt: 220 °C (48 mmHg)
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Powder or crystals
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR