(±)-γ-Octalactone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one

12940-0250 12940-1000
TS12940-0250EA 33 USD
TS12940-0250 TS12940-1000
(±)-γ-Octalactone 98%
(±)-γ-Octalactone
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 234 °C (1013 hPa)
Density: 0.98 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00005402
CAS Number: 104-50-7
EINECS: 203-208-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4470 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR