β-Naphthoflavone 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,6-Benzoflavone, 3-Phenyl-1H-naphtho[2,1-b]pyran-1-one, 3-Phenyl-1H-benzo[f]chromen-1-one

12817-0010 12817-0050
TS12817-0010EA 44 USD
TS12817-0010 TS12817-0050
β-Naphthoflavone 99+%
β-Naphthoflavone
Formula: C₁₉H₁₂O₂
MW: 272.3 g/mol
Boiling Pt: 461 °C (1013 hPa)
Melting Pt: 162…164 °C
MDL Number: MFCD00004986
CAS Number: 6051-87-2
EINECS: 227-958-4

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 162 to 166 °C
HPLC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR