5-Methylthiophene-2-carboxaldehyde 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde
12787-0100
12787-0500
TS12787-0100EA
66
USD
TS12787-0100
TS12787-0500
5-Methylthiophene-2-carboxaldehyde 98%
5-Methylthiophene-2-carboxaldehyde
Formula:
C₆H₆OS Density: 1.18 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005434 CAS Number: 13679-70-4 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5800 to 1.5840 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...