3-Methylthiophene 99+%

Supplier: Thermo Scientific Chemicals
Danger

12786-0500 12786-2500
TS12786-0500EA 67 USD
TS12786-0500 TS12786-2500
3-Methylthiophene 99+%
3-Methylthiophene
Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 115…117 °C (760 mmHg)
Melting Pt: –69 °C
Density: 1.016 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005470
CAS Number: 616-44-4
EINECS: 210-482-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5210 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR