2-Methylthiophene 98%

Supplier: Thermo Scientific Chemicals
Danger

12785-0250 12785-1000
TS12785-0250EA 46 USD
TS12785-0250 TS12785-1000
2-Methylthiophene 98%
2-Methylthiophene
Formula: C₅H₆S
Boiling Pt: 113 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.02 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5210 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR