1-Methylpyrrole 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methylpyrrole, 1-Methyl-1H-pyrrole

12760-1000 12760-5000
TS12760-1000EA 70 USD
TS12760-1000 TS12760-5000
1-Methylpyrrole 99+%
1-Methylpyrrole
Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.91 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00005345
CAS Number: 96-54-8
EINECS: 202-513-7
UN: 1992

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4890 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR