N-Methylmaleimide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Methyl-1H-pyrrole-2,5-dione
12708-0050
12708-0250
12708-1000
TS12708-0050EA
69
USD
TS12708-0050
TS12708-0250
TS12708-1000
N-Methylmaleimide 97%
N-Methylmaleimide
Formula:
C₅H₅NO₂ MW: 111.1 g/mol Boiling Pt: 194 °C (1013 hPa) Melting Pt: 93…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005508 CAS Number: 930-88-1 EINECS: 213-226-1 UN: 2928 ADR: 6.1,II |
Specification Test Results
Appearance | Light yellow Crystals |
Infrared spectrum | Conforms |
Melting point | 93 to 98 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...