3-Methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Skatol, 3-Methyl-1H-indole, β-Methylindole, Skatole

12703-0100 12703-0250 12703-1000
TS12703-0100EA 54 USD
TS12703-0100 TS12703-0250 TS12703-1000
3-Methylindole 98%
3-Methylindole
Formula: C₉H₉N
Boiling Pt: 265…266 °C (755 mmHg)
Melting Pt: 95…98 °C
Flash Pt: 132 °C
MDL Number: MFCD00005627
CAS Number: 83-34-1
EINECS: 201-471-7
UN: 3077
Merck Index: 12,08700

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 95 to 98 °C
GC ≥97.5 %
Color scale ≤50 APHA
Impurity ≤1.1 % Tosyl chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR