2-Methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-1H-indole

12702-1000 12702-5000
TS12702-1000EA 90 USD
TS12702-1000 TS12702-5000
2-Methylindole 98%
2-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 273 °C (760 mmHg)
Melting Pt: 56…60 °C
Flash Pt: 141 °C
Storage Temperature: Freezer
MDL Number: MFCD00005616
CAS Number: 95-20-5
EINECS: 202-398-3
UN: 3335
ADR: 9 III

Order Now

Specification Test Results

Appearance Yellow to red-purple to brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 56 to 60 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR