3-Methylglutaric anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyldihydro-2H-pyran-2,6(3H)-dione

12687-0050 12687-0250
TS12687-0050EA 55 USD
TS12687-0050 TS12687-0250
3-Methylglutaric anhydride 97%
3-Methylglutaric anhydride
Formula: C₆H₈O₃
Boiling Pt: 180…182 °C (25 mmHg)
Melting Pt: 40…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006683
CAS Number: 4166-53-4
EINECS: 224-020-6

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 40 to 45 °C
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR