2-Methylbenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole

12633-0050 12633-1000
TS12633-0050EA 29 USD
TS12633-0050 TS12633-1000
2-Methylbenzimidazole 98%
2-Methylbenzimidazole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 175…179 °C
MDL Number: MFCD00005598
CAS Number: 615-15-6
EINECS: 210-411-9

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 175 to 179 °C
HPLC ≥97.5 %
Residue after evaporation ≤0.001 %
Acidity ≤0.0002 meq/g
Substance darkened by H2SO4 Passes test
Iron (Fe) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR