2-Mercapto-2-thiazoline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
12556-0050
12556-1000
12556-5000
TS12556-0050EA
32
USD
TS12556-0050
TS12556-1000
TS12556-5000
2-Mercapto-2-thiazoline 98%
2-Mercapto-2-thiazoline
Formula:
C₃H₅NS₂ MW: 119.21 g/mol Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126013 CAS Number: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 106 °C |
Titration with TBAH | 97.5 to 102.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...