2-Pyridinethiol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine-2-thiol, 2-Mercaptopyridine

12554-0025 12554-0250 12554-1000
TS12554-0025EA 22 USD
TS12554-0025 TS12554-0250 TS12554-1000
2-Pyridinethiol 98%
2-Pyridinethiol
Formula: C₅H₅NS
MW: 111.17 g/mol
Melting Pt: 126…130 °C
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006285
CAS Number: 2637-34-5
EINECS: 220-131-9

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
Titration Iodimetric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR