2-Mercaptobenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

12544-0010 12544-0050 12544-1000 12544-2500
TS12544-0010EA 130 USD
TS12544-0010 TS12544-0050 TS12544-1000 TS12544-2500
2-Mercaptobenzimidazole 98%
2-Mercaptobenzimidazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 302…307 °C
Flash Pt: >250 °C
MDL Number: MFCD00466107
CAS Number: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Ash ≤0.3 %
Titration with NaOH ≥97.5 %
Insoluble matter ≤0.3 % (in NaOH)
Volatile matter ≤0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR