3,5-Lutidine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Dimethylpyridine

12521-1000 12521-5000
TS12521-1000EA 50 USD
TS12521-1000 TS12521-5000
3,5-Lutidine 98+%
3,5-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 172.7 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.93 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00006404
CAS Number: 591-22-0
EINECS: 209-708-6
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
GC ≥98.0 %
Sulfate (SO4) ≤0.05 %
Iron (Fe) ≤0.005 %
Acidity (HCl) ≤0.025 %
Insoluble matter ≤0.025 % (in water and ppt by NH4OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR