2,4-Lutidine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4-Dimethylpyridine

12518-1000 12518-5000
TS12518-1000EA 116 USD
TS12518-1000 TS12518-5000
2,4-Lutidine 98+%
2,4-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.92 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00006337
CAS Number: 108-47-4
EINECS: 203-586-8
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5010 (20°C, 589 nm)
GC ≥98.0 %
Total volume capacity ≥1.10 eq/l (OH) as is

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR