1-(4-Nitrophenyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Warning

12494-0050 12494-0250 12494-1000
TS12494-0050EA 32 USD
TS12494-0050 TS12494-0250 TS12494-1000
1-(4-Nitrophenyl)piperazine 98%
1-(4-Nitrophenyl)piperazine
Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00005961
CAS Number: 6269-89-2
EINECS: 228-443-7

Order Now

Specification Test Results

Appearance Yellow to ochre Powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR