8-Aminoquinoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-8-ylamine

12371-0050 12371-0100 12371-0500
TS12371-0050EA 67 USD
TS12371-0050 TS12371-0100 TS12371-0500
8-Aminoquinoline 98+%
8-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 174 °C (26 mmHg)
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to green to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 65 °C
GC ≥98.0 %
Titration with HClO4 ≥98.0 %
Aluminium (Al) ≤0.000005 %
Barium (Ba) ≤0.000002 %
Bismuth (Bi) ≤0.00001 %
Cadmium (Cd) ≤0.000001 %
Calcium (Ca) ≤0.00005 %
Chromium (Cr) ≤0.000002 %
Cobalt (Co) ≤0.000001 %
Copper (Cu) ≤0.000002 %
Iron (Fe) ≤0.00002 %
Lead (Pb) ≤0.000002 %
Lithium (Li) ≤0.000001 %
Magnesium (Mg) ≤0.00001 %
Manganese (Mn) ≤0.000001 %
Molybdenum (Mo) ≤0.000002 %
Nickel (Ni) ≤0.00002 %
Potassium (K) ≤0.00001 %
Sodium (Na) ≤0.00005 %
Titanium (Ti) ≤0.00001 %
Vanadium (V) ≤0.000001 %
Zirconium (Zr) ≤0.00001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR