Isophthalic acid 99%

Supplier: Thermo Scientific Chemicals

12265-0020 12265-0050 12265-5000
TS12265-0020EA 83 USD
TS12265-0020 TS12265-0050 TS12265-5000
Isophthalic acid 99%
Isophthalic acid
Formula: C₈H₆O₄
Melting Pt: 341…343 °C
Storage Temperature: Ambient
MDL Number: MFCD00002516
CAS Number: 121-91-5
EINECS: 204-506-4
Merck Index: 13,05216

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.0015 %
Titration with NaOH ≥98.5 %
Cobalt (Co) ≤2 ppm
Iron (Fe) ≤2 ppm
Manganese (Mn) ≤2 ppm
Impurity ≤0.0025 % 3-Carboxy benzaldehyde


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR