Isonipecotamide 98%

Supplier: Thermo Scientific Chemicals
Warning

12261-0250 12261-1000 12261-5000
TS12261-0250EA 38 USD
TS12261-0250 TS12261-1000 TS12261-5000
Isonipecotamide 98%
Isonipecotamide
Formula: C₆H₁₂N₂O
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00038012
CAS Number: 39546-32-2
EINECS: 254-501-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 150 °C (on dry substance)
Water ≤2 % (K.F.)
Loss on drying ≤2 % (70 °C)
Sulfated ash ≤0.2 %
Titration with HClO4 ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR