3-(Hydroxymethyl)indole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

12219-0050 12219-0250
TS12219-0050EA 76 USD
TS12219-0050 TS12219-0250
3-(Hydroxymethyl)indole 97%
3-(Hydroxymethyl)indole
Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005632
CAS Number: 700-06-1

Order Now

Specification Test Results

Appearance Off-white to yellow to light orange Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 92 to 100 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR