Iminodibenzyl 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 10,11-Dihydro-5H-dibenzo[b,f]azepine

12207-0250 12207-1000
TS12207-0250EA 29 USD
TS12207-0250 TS12207-1000
Iminodibenzyl 97%
Iminodibenzyl
Formula: C₁₄H₁₃N
MW: 195.27 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005070
CAS Number: 494-19-9
EINECS: 207-787-1

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 105 to 108 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.3 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR