2-Imidazolidinone hemihydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Imidazolidone hemihydrate
12204-0025
12204-0100
12204-0250
12204-5000
TS12204-0025EA
230
USD
TS12204-0025
TS12204-0100
TS12204-0250
TS12204-5000
2-Imidazolidinone hemihydrate 99+%
2-Imidazolidinone hemihydrate
Formula:
C₃H₆N₂O·0.5H₂O MW: 95.10 (86.09 anhy) Boiling Pt: >100 °C (760 mmHg) Melting Pt: 58 °C Flash Pt: 265 °C Storage Temperature: Ambient |
MDL Number:
MFCD02662355 CAS Number: 121325-67-5 EINECS: 204-436-4 |
Specification Test Results
Appearance | White to light yellow Adhering crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤13 % |
GC | ≥99.0 % |
Elemental Analysis | 5.1 to 5.6 % N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...