3-Pyridone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol

12185-1000 12185-5000
TS12185-1000EA 51 USD
TS12185-1000 TS12185-5000
3-Pyridone 98%
3-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 180 °C (22 mmHg)
Melting Pt: 123…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4
UN: 2811

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 130 °C
Water ≤0.2 %
GC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR