1-(2-Hydroxyethyl)piperazine 98.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol

12119-1000 12119-5000
TS12119-1000EA 37 USD
TS12119-1000 TS12119-5000
1-(2-Hydroxyethyl)piperazine 98.5%
1-(2-Hydroxyethyl)piperazine
Formula: C₆H₁₄N₂O
Boiling Pt: ∼246 °C (1013 hPa)
Melting Pt: –38.5 °C
Density: 1.06 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5075 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR