Glutaric anhydride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
11997-0025
11997-0050
11997-1000
11997-5000
TS11997-0025EA
1003
USD
TS11997-0025
TS11997-0050
TS11997-1000
TS11997-5000
Glutaric anhydride 95%
Glutaric anhydride
Formula:
C₅H₆O₃ Boiling Pt: 290 °C (760 mmHg) Melting Pt: 46…57 °C Flash Pt: 173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006679 CAS Number: 108-55-4 EINECS: 203-593-6 |
Specification Test Results
Appearance | White to yellow to light beige Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
GC | ≥94 % |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...