2-Furoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid

11982-0025 11982-0050 11982-1000 11982-5000
TS11982-0025EA 575 USD
TS11982-0025 TS11982-0050 TS11982-1000 TS11982-5000
2-Furoic acid 98%
2-Furoic acid
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 230…232 °C (760 mmHg)
Melting Pt: 129…133 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00003238
CAS Number: 88-14-2
EINECS: 201-803-0

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 129 to 133 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR