Furfuryl mercaptan 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Furanmethanethiol, 2-Furanmethanthiol, 2-Furylmethanethiol

11981-0250 11981-1000
TS11981-0250EA 51 USD
TS11981-0250 TS11981-1000
Furfuryl mercaptan 97%
Furfuryl mercaptan
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.13 g/cm³
Flash Pt: 45 °C
MDL Number: MFCD00003254
CAS Number: 98-02-2
EINECS: 202-628-2
UN: 1228

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5330 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR