2-Fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

11963-0050 11963-0500
TS11963-0050EA 29 USD
TS11963-0050 TS11963-0500
2-Fluoropyridine 98%
2-Fluoropyridine
Formula: C₅H₄FN
Boiling Pt: 126 °C (753 mmHg)
Density: 1.12 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006224
CAS Number: 372-48-5
EINECS: 206-757-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4690 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR