Ethyl picolinate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate
11886-0250
11886-1000
TS11886-0250EA
63
USD
TS11886-0250
TS11886-1000
Ethyl picolinate 99%
Ethyl picolinate
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Melting Pt: 2 °C Density: 1.11 g/cm³ Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 EINECS: 219-758-0 |
Specification Test Results
Appearance | Clear yellow to pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5090 to 1.5110 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...