2,5-Dimethylthiophene 98.5%

Supplier: Thermo Scientific Chemicals
Warning

11688-0050 11688-0250 11688-1000
TS11688-0050EA 40 USD
TS11688-0050 TS11688-0250 TS11688-1000
2,5-Dimethylthiophene 98.5%
2,5-Dimethylthiophene
Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 134…136 °C (760 mmHg)
Melting Pt: –63 °C
Density: 0.984 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00005452
CAS Number: 638-02-8
EINECS: 211-313-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5140 (20°C, 589 nm)
GC ≥98.0 %
Nitrite (NO2) ≤10 ppm
Sulfate (SO4) ≤20 ppm
Barium (Ba) ≤50 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤500 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR