2,5-Dimethylpyrrole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Dimethyl-1H-pyrrole

11675-0050 11675-0250
TS11675-0050EA 50 USD
TS11675-0050 TS11675-0250
2,5-Dimethylpyrrole 97%
2,5-Dimethylpyrrole
Formula: C₆H₉N
MW: 95.1424 g/mol
Boiling Pt: 165 °C (740 mmHg)
Density: 0.935 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00005223
CAS Number: 625-84-3
EINECS: 210-913-8
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5080 (20°C, 589 nm)
GC ≥96.0 %
Ammonium (NH4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.05 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR