5,6-Dimethylbenzimidazole 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,6-Dimethyl-1H-benzimidazole, 5,6-Dimethyl-1H-benzo[d]imidazole

11598-0250 11598-1000
TS11598-0250EA 46 USD
TS11598-0250 TS11598-1000
5,6-Dimethylbenzimidazole 99+%
5,6-Dimethylbenzimidazole
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 203…206 °C
MDL Number: MFCD00005603
CAS Number: 582-60-5
EINECS: 209-488-1
Merck Index: 13,03263

Order Now

Specification Test Results

Appearance Light beige Powder
Infrared spectrum Conforms
Melting point 203 to 206 °C
Titration with HClO4 ≥99.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤5000 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR