2,5-Dimethoxytetrahydrofuran cis- and trans-mixture 99%

Supplier: Thermo Scientific Chemicals
Danger

11567-1000 11567-5000
TS11567-1000EA 56 USD
TS11567-1000 TS11567-5000
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture 99%
2,5-Dimethoxytetrahydrofuran
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.021 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005359
CAS Number: 696-59-3
EINECS: 211-797-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4190 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR