Xanthurenic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

11509-0010 11509-0050
TS11509-0010EA 54 USD
TS11509-0010 TS11509-0050
Xanthurenic acid 96%
Xanthurenic acid
Formula: C₁₀H₇NO₄
Melting Pt: 297…298 °C
Storage Temperature: Ambient
MDL Number: MFCD00006754
CAS Number: 59-00-7
EINECS: 200-410-1
Merck Index: 12,10202

Order Now

Specification Test Results

Appearance Ochre Powder
Infrared spectrum Conforms
Titration excess NaOH ≥95.0 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR