4,6-Dihydroxypyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Hydroxy-1H-pyrimidin-4-one

11508-0250 11508-1000 11508-5000
TS11508-0250EA 50 USD
TS11508-0250 TS11508-1000 TS11508-5000
4,6-Dihydroxypyrimidine 98%
4,6-Dihydroxypyrimidine
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016733
CAS Number: 1193-24-4
EINECS: 214-772-3

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR