7-(2,3-Dihydroxypropyl)theophylline 99%

Supplier: Thermo Scientific Chemicals
Warning

11505-1000 11505-5000
TS11505-1000EA 74 USD
TS11505-1000 TS11505-5000
7-(2,3-Dihydroxypropyl)theophylline 99%
7-(2,3-Dihydroxypropyl)theophylline
Formula: C₁₀H₁₄N₄O₄
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005759
CAS Number: 479-18-5
EINECS: 207-526-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
Titration with HClO4 ≥98.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR