2,3-Dichloropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

11372-0100 11372-0500
TS11372-0100EA 49 USD
TS11372-0100 TS11372-0500
2,3-Dichloropyridine 99%
2,3-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 202.5…203.5 °C (1013 hPa)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00006229
CAS Number: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥98.5 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR