2,5-Dibromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

11290-0100 11290-0500
TS11290-0100EA 37 USD
TS11290-0100 TS11290-0500
2,5-Dibromopyridine 97%
2,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 234…235 °C (1013 hPa)
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6

Order Now

Specification Test Results

Appearance White to beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 91 to 96 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR