3,5-Diamino-1H-1,2,4-triazole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,5-Diamino-1,2,4-triazole, Guanazole

11244-0250 11244-1000
TS11244-0050EA 40.17 USD
TS11244-0050 TS11244-0250 TS11244-1000
3,5-Diamino-1H-1,2,4-triazole 98%
3,5-Diamino-1H-1,2,4-triazole
Formula: C₂H₅N₅
MW: 99.1 g/mol
Melting Pt: 204…208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005233
CAS Number: 1455-77-2
EINECS: 215-937-2

Order Now

Specification Test Results

Appearance White to beige to grey Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 202 to 207 °C
Titration with HClO4 ≥97.5 %
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR