Pyridine-2-carbonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyanopyridine, 2-Pyridinecarbonitrile, Picolinonitrile

11085-0050 11085-1000 11085-5000
TS11085-0050EA 25 USD
TS11085-0050 TS11085-1000 TS11085-5000
Pyridine-2-carbonitrile 99%
Pyridine-2-carbonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 212
Melting Pt: 26…28 °C
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00006218
CAS Number: 100-70-9
EINECS: 202-880-3
UN: 3439

Order Now

Specification Test Results

Appearance White to brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 31 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR