Indole-5-carbonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

11079-0010 11079-0050
TS11079-0010EA 21 USD
TS11079-0010 TS11079-0050
Indole-5-carbonitrile 99%
Indole-5-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 105 to 108 °C
GC ≥98.5 %
Titrable acid ≤0.001 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR