Eucalyptol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane

11034-0050 11034-1000 11034-5000
TS11034-0050EA 26 USD
TS11034-0050 TS11034-1000 TS11034-5000
Eucalyptol 99%
Eucalyptol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 176…177 °C (760 mmHg)
Melting Pt: 1.5 °C
Density: 0.9225 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00167977
CAS Number: 470-82-6
EINECS: 207-431-5
UN: 3271
ADR: 3,III
Merck Index: 12,03940

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4562 to 1.4582 (20°C, 589 nm)
Solubility (in ethanol (70% v/v)) 1:2
GC ≥98.5 %
Specific gravity (20°C) 0.921 to 0.924
Specific optical rotation -0.3° to +0.3° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR