2-Chloroquinoline 99%

Supplier: Thermo Scientific Chemicals
Warning

11004-0010 11004-0050 11004-0250
TS11004-0010EA 44 USD
TS11004-0010 TS11004-0050 TS11004-0250
2-Chloroquinoline 99%
2-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 265…267 °C (760 mmHg)
Melting Pt: 34…37 °C
Density: 1.23 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006741
CAS Number: 612-62-4
EINECS: 210-317-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals and/or chunks or fused mass
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR