2-Chloropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11001-1000
11001-5000
TS11001-1000EA
44
USD
TS11001-1000
TS11001-5000
2-Chloropyridine 99%
2-Chloropyridine
Formula:
C₅H₄ClN Boiling Pt: 168…170 °C (760 mmHg) Melting Pt: –46 °C Density: 1.209 g/cm³ Flash Pt: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006228 CAS Number: 109-09-1 EINECS: 203-646-3 UN: 2822 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5310 to 1.5330 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥98.5 % |
Visual transition interval | from pH 1.2 (red) to 2.3 (yellow) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...