2-Chloro-5-nitropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

10974-0250 10974-1000
TS10974-0250EA 108 USD
TS10974-0250 TS10974-1000
2-Chloro-5-nitropyridine 99%
2-Chloro-5-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 107
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 106 to 110 °C
GC ≥98.5 %
Titrable free acid ≤0.005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR