2-Chloro-3-nitropyridine 99+%

Supplier: Thermo Scientific Chemicals
Warning

10973-0250 10973-1000
TS10973-0250EA 98 USD
TS10973-0250 TS10973-1000
2-Chloro-3-nitropyridine 99+%
2-Chloro-3-nitropyridine
Formula: C₅H₃ClN₂O₂
Melting Pt: 100…103 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 103 °C
HPLC ≥99.0 %
Sulfate (SO4) ≤0.002 %
Ammonium (NH4) ≤0.002 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR