1-(2-Chloroethyl)pyrrolidine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

10922-0250 10922-1000
TS10922-0250EA 53 USD
TS10922-0250 TS10922-1000
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride
Formula: C₆H₁₂ClN
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012718
CAS Number: 7250-67-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 175 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR