Carbazole 96%

Supplier: Thermo Scientific Chemicals
Warning

10826-0010 10826-0050 10826-0250 10826-2500
TS10826-0010EA 330 USD
TS10826-0010 TS10826-0050 TS10826-0250 TS10826-2500
Carbazole 96%
Carbazole
Formula: C₁₂H₉N
MW: 167.21 g/mol
Boiling Pt: 355 °C (760 mmHg)
Melting Pt: 240…246 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 240 to 246 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR